Learn More
Biomatik Corporation Rat PLAU/uPA (Urokinase-Type Plasminogen Activator) ELISA Kit
Supplier:  Biomatik Corporation EKF58602
Encompass_Preferred
Rat PLAU/uPA (Urokinase-Type Plasminogen Activator) ELISA Kit for detecting PLAU; uPA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P29598. Target Synonyms: PLAU; uPA; PLAU; Urokinase; ATF; plasminogen activator; urokinase; UPA; u-PA; U-plasminogen activator; urinary; urokinase-type plasminogen activator. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5900