Learn More
Biomatik Corporation Rat Rantes (Regulated On Activation, Normal T-Cell Expressed and Secreted) ELISA Kit
Supplier:  Biomatik Corporation EKF58606
Encompass_Preferred
Rat Rantes (Regulated On Activation, Normal T-Cell Expressed and Secreted) ELISA Kit for detecting Rantes in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P50231. Target Synonyms: Rantes; Regulated On Activation; Normal T-Cell Expressed and Secreted; CCL5; SCYA5; SISd. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5902