Learn More
Biomatik Corporation Rat Rbp1 (Retinol-binding protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58608
Encompass_Preferred
Rat Rbp1 (Retinol-binding protein 1) ELISA Kit for detecting Rbp1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02696. Target Synonyms: Rbp1; Cellular retinol-binding protein; CRBP; Cellular retinol-binding protein I; CRBP-I;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5904