Learn More
Biomatik Corporation Rat Cyp7a1 (Cholesterol 7-alpha-monooxygenase) ELISA Kit
Supplier:  Biomatik Corporation EKF58615
Encompass_Preferred
Rat Cyp7a1 (Cholesterol 7-alpha-monooxygenase) ELISA Kit for detecting Cyp7a1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P18125. Target Synonyms: Cyp7a1; Cholesterol 7-alpha-monooxygenase; Cytochrome P450 7A1; CYPVII; Cholesterol 7-alpha-hydroxylase;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5910