Learn More
Biomatik Corporation Rat Atf2 (Cyclic AMP-dependent transcription factor ATF-2) ELISA Kit
Supplier:  Biomatik Corporation EKF58624
Encompass_Preferred
Rat Atf2 (Cyclic AMP-dependent transcription factor ATF-2) ELISA Kit for detecting Atf2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q00969. Target Synonyms: Atf2; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; glial cell line-derived neurotrophic factor; HFB1-GDNF; hGDNF; HSCR3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5917