Learn More
Biomatik Corporation Rat DPPⅣ (Dipeptidyl Peptldase Ⅳ) ELISA Kit
Supplier:  Biomatik Corporation EKF58634
Encompass_Preferred
Rat DPPⅣ (Dipeptidyl Peptldase Ⅳ) ELISA Kit for detecting DPPⅣ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14740. Target Synonyms: Dpp4; CD26; ADABP; ADCP-2; ADCP2DPP IV; Adenosine deaminase complexing protein 2TP103; CD26 antigen; CD26T-cell activation antigen CD26; dipeptidyl peptidase 4; Dipeptidyl peptidase IV; dipeptidylpeptidase 4; dipeptidylpeptidase IV; CD26; adenosine deaminase complexing protein 2; DPPIV; DPPⅣ. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5924