Learn More
Biomatik Corporation Rat Lipe (Hormone-sensitive lipase) ELISA Kit
Supplier:  Biomatik Corporation EKF58645
Encompass_Preferred
Rat Lipe (Hormone-sensitive lipase) ELISA Kit for detecting Lipe in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15304. Target Synonyms: Lipe; HSL; LHS; LIPE; hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSLEC 3.1.1.79; LHS; lipase; hormone-sensitive; LIPE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5934