Learn More
Biomatik Corporation Rat NF-κB p65 (Nuclear Factor-κB P65) ELISA Kit
Supplier:  Biomatik Corporation EKF58655
Encompass_Preferred
Rat NF-κB p65 (Nuclear Factor-κB P65) ELISA Kit for detecting NF-κB p65 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q7TQN4. Target Synonyms: NF-κB p65; NFKB3; p65RelA; NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A; nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3; p65; Nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65; p65; v-rel reticuloendotheliosis viral oncogene homolog A; avian; v-rel reticuloendotheliosis viral oncogene homolog A; nuclear factor of kappalight polypeptide gene enhancer in B-cells 3; p65. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5941