Learn More
Biomatik Corporation Human EPCA2 (Early Prostate Cancer Antigen 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58661
Encompass_Preferred
Human EPCA2 (Early Prostate Cancer Antigen 2) ELISA Kit for detecting EPCA2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 3.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: EPCA2. Target Synonyms: EPCA2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5946