Learn More
Biomatik Corporation Human THBS4 (Thrombospondin-4) ELISA Kit
Supplier:  Biomatik Corporation EKF58677
Encompass_Preferred
Human THBS4 (Thrombospondin-4) ELISA Kit for detecting THBS4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35443. Target Synonyms: TSP-4; Thrombospondin-4; THBS4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5958