Learn More
Biomatik Corporation Human KISS1 (Metastasis-suppressor KiSS-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58680
Encompass_Preferred
Human KISS1 (Metastasis-suppressor KiSS-1) ELISA Kit for detecting KISS1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q15726. Target Synonyms: KISS1; Metastasis-suppressor KiSS-1; Kisspeptin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5960
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'