Learn More
Biomatik Corporation Human AVPR1A (Vasopressin V1a receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF58688
Encompass_Preferred
Human AVPR1A (Vasopressin V1a receptor) ELISA Kit for detecting AVPR1A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P37288. Target Synonyms: AVPR1A; AVPR V1a; V1aR; Antidiuretic hormone receptor 1a; arginine vasopressin receptor 1A; AVPR V1a; AVPR1; SCCL vasopressin subtype 1a receptor; V1a vasopressin receptor; V1aR; V1-vascular vasopressin receptor AVPR1A; Vascular; hepatic-type arginine vasopressin receptor; vasopressin V1a receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5966