Learn More
Biomatik Corporation Human KPNA2 (Importin subunit alpha-2) ELISA Kit
Supplier:  Biomatik Corporation EKF58704
Encompass_Preferred
Human KPNA2 (Importin subunit alpha-2) ELISA Kit for detecting KPNA2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P52292. Target Synonyms: KPNA2; Importin subunit alpha-2; Importin alpha 1; IPOA1; KPNA2; Pendulin; QIP2; RCH1; SRP1-alpha; importin-alpha-P1; karyopherin alpha 2; RAG cohort 1; importin alpha 1; Karyopherin subunit alpha-2; importin alpha 2; RAG cohort 1; RAG cohort protein 1; importin subunit alpha-2; SRP1; SRP1alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5978