Learn More
Biomatik Corporation Human APOA4 (Apolipoprotein A-IV) ELISA Kit
Supplier:  Biomatik Corporation EKF58708
Encompass_Preferred
Human APOA4 (Apolipoprotein A-IV) ELISA Kit for detecting APOA4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800ng/ml. Sensitivity: 7.5ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06727. Target Synonyms: ApoA4; Apolipoprotein A4; Apolipoprotein A-IV; Apo-AIV; ApoA-IV; apolipoprotein A-IV; MGC142154; MGC142156. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5982