Learn More
Biomatik Corporation Human KNG1 (Kininogen-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58710
Encompass_Preferred
Human KNG1 (Kininogen-1) ELISA Kit for detecting KNG1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.391-25ng/ml. Sensitivity: 0.234ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01042. Target Synonyms: KNG1; Kininogen 1; Alpha-2-thiol proteinase inhibitor; BDK; bradykinin; Fitzgerald factor; High molecular weight kininogen; kininogen; kininogen-1; KNGHMWK; Williams-Fitzgerald-Flaujeac factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5984