Learn More
Biomatik Corporation Human CRYAB (Alpha-crystallin B chain) ELISA Kit
Supplier:  Biomatik Corporation EKF58713
Encompass_Preferred
Human CRYAB (Alpha-crystallin B chain) ELISA Kit for detecting CRYAB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02511. Target Synonyms: CRYαB; Crystallin Alpha B; CRYAB; CRYA2; CTPP2; HSPB5; alpha B crystallin; alpha; B-crystallin; alpha-crystallin B chain; alpha crystallin B chain; Heat shock protein beta-5; heat-shock 20 kD like-protein; Renal carcinoma antigen NY-REN-27; Rosenthal fiber component. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5987