Learn More
Biomatik Corporation Rat Anxa5 (Annexin A5) ELISA Kit
Supplier:  Biomatik Corporation EKF58721
Encompass_Preferred
Rat Anxa5 (Annexin A5) ELISA Kit for detecting Anxa5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14668. Target Synonyms: Anxa5; Lipocortin V; Endonexin II; Calphobindin I; CBP-I; Annexin-5; Thromboplastin inhibitor; Placental anticoagulant protein I; PAP-I; Placental anticoagulant protein 4; PP4; Vascular anticoagulant-alpha; VAC-alpha; Anchorin CII; Annexin V; ANX5; ANXA5; ANXV; Anchorin CII; annexin A5; Annexin V; annexin-5; Annexin-5; ANX5; Calphobindin I; Endonexin II; ENX2CBP-I; Lipocortin V; PAP-I; Placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5995