Learn More
Biomatik Corporation Human AURKA (Aurora kinase A) ELISA Kit
Supplier:  Biomatik Corporation EKF58741
Encompass_Preferred
Human AURKA (Aurora kinase A) ELISA Kit for detecting AURKA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O14965. Target Synonyms: AIK; ARK 1; ARK1; AURA; AURKA; aurora kinase A; Aurora related kinase 1; Aurora; IPL1 related kinase 1; AURORA2; Breast tumor amplified kinase; BTAK; hARK1; STK15; STK6; STK7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6012