Learn More
Biomatik Corporation Human TDO (Tryptophan-2,3-dioxygenase) ELISA Kit
Supplier:  Biomatik Corporation EKF58745
Encompass_Preferred
Human TDO (Tryptophan-2,3-dioxygenase) ELISA Kit for detecting TDO in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48775. Target Synonyms: Tryptophan-2; 3-dioxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6016