Learn More
Biomatik Corporation Human Sulfotransferase 1A1 (Sult1A1) Elisa Kit
Supplier:  Biomatik Corporation EKC42655
Encompass_Preferred
Human Sulfotransferase 1A1 (Sult1A1) Elisa Kit for detecting Sulfotransferase Family, Cytosolic, 1A, Phenol-Preferring, Member 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P50225; SULT1A1. Target Synonyms: Aryl sulfotransferase 1; HAST1/HAST2; Human aryl sulfotransferase mRNA complete cds; MGC131921; MGC5163; OTTHUMP00000162568; OTTHUMP00000162569; P PST 1; P PST; P-PST 1; Phenol sulfating phenol sulfotransferase 1; Phenol sulfotransferase 1; Phenol-sulfating phenol sulfotransferase 1; PST; ST1A1; ST1A1_HUMAN; ST1A3; STP; STP1; Sulfotransferase 1A1; Sulfotransferase family 1A phenol preferring member 1; Sulfotransferase family cytosolic 1A phenol preferring member 1; Sulfotransferase phenol preferring 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5161