Learn More
Biomatik Corporation Sheep Transforming growth factor beta-1(TGFB1) ELISA kit
Supplier:  Biomatik Corporation EKC42662
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Ovis aries (Sheep). Detection Range: 15.6 ng/mL-1000 ng/mL. Sensitivity: 3.9 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P50414. Target Synonyms: CED, DPD1, LAP, TGFB, TGFbeta, TGF-beta 1 protein|latency-associated peptide. Restriction: For Research Use Only.
Catalog No. 50-229-5165