Learn More
Biomatik Corporation Human Talin-1 (Tln1) Elisa Kit
Supplier:  Biomatik Corporation EKC42672
Encompass_Preferred
Human Talin-1 (Tln1) Elisa Kit for detecting Talin 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y490; TLN1. Target Synonyms: ILWEQ; Talin 1; Talin; Talin-1; TLN 1; TLN; Tln1; TLN1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5169