Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily Member 9 (Tnfrsf9) Elisa Kit
Supplier:  Biomatik Corporation EKC42684
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily Member 9 (Tnfrsf9) Elisa Kit for detecting Tumor Necrosis Factor Receptor Superfamily, Member 9 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q07011; TNFRSF9. Target Synonyms: 4 1BB; 4 1BB ligand receptor; 4-1BB ligand receptor; 4-1BB Ligand Receptor T Cell; 4-1BB; mouse; homolog of; Antigen 4-1BB Homolog; CD 137; CD137; CD137 antigen; CDw137; HLDA VI; Homolog of mouse 4 1BB; ILA; induced by lymphocyte activation (ILA); Induced by lymphocyte activation; Interleukin activated receptor homolog of mouse Ly63; Ly63; mouse; homolog of; MGC2172; OTTHUMP00000044294; Receptor protein 4 1BB; T cell antigen 4 1BB homolog; T cell antigen ILA; T-cell antigen 4-1BB homolog; T-cell antigen ILA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5174