Learn More
Biomatik Corporation Rat Tumor necrosis factor ligand superfamily member 15(TNFSF15) ELISA kit
Supplier:  Biomatik Corporation EKC42687
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 25 pg/mL-1600 pg/mL. Sensitivity: 6.25 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8K3Y7. Target Synonyms: MGC129934, MGC129935, TL1, TL1A, VEGI, VEGI192A, TNF ligand-related molecule 1|TNF superfamily ligand TL1A|vascular endothelial cell growth inhibitor|vascular endothelial growth inhibitor-192A. Restriction: For Research Use Only.
Catalog No. 50-229-5177