Learn More
Biomatik Corporation Rat Serine/Threonine-Protein Kinase Tnni3K (Tnni3K) Elisa Kit
Supplier:  Biomatik Corporation EKC42692
Encompass_Preferred
Rat Serine/Threonine-Protein Kinase Tnni3K (Tnni3K) Elisa Kit for detecting Tnni3 Interacting Kinase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.69pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q7TQP6; Tnni3k. Target Synonyms: Tnni3k; Cark; Serine/threonine-protein kinase TNNI3K; EC 2.7.11.1; Cardiac ankyrin repeat kinase; TNNI3-interacting kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5181