Learn More
Biomatik Corporation Mouse Transthyretin(TTR) ELISA kit
Supplier:  Biomatik Corporation EKC42701
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07309. Target Synonyms: HsT2651, PALB, TBPA, prealbumin, amyloidosis type I|thyroxine-binding prealbumin. Restriction: For Research Use Only.
Catalog No. 50-229-5188