Learn More
Biomatik Corporation Human Protein Wnt-2B (Wnt2B) Elisa Kit
Supplier:  Biomatik Corporation EKC42712
Encompass_Preferred
Human Protein Wnt-2B (Wnt2B) Elisa Kit for detecting Wingless-Type Mmtv Integration Site Family, Member 2B in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q93097; WNT2B. Target Synonyms: WNT2B; WNT13; Protein Wnt-2b; Protein Wnt-13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5192