Learn More
Biomatik Corporation Human Selenoprotein S(SELS) ELISA kit
Supplier:  Biomatik Corporation EKC42727
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates, cell lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9BQE4. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5200