Learn More
Biomatik Corporation Rat Keratin, type I cytoskeletal 18(KRT18) ELISA kit
Supplier:  Biomatik Corporation EKC42738
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.94 ng/mL-60 ng/mL. Sensitivity: 0.23 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q5BJY9. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5207