Learn More
Biomatik Corporation Bovine arachidonic Acid(AA) ELISA kit
Supplier:  Biomatik Corporation EKC42748
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Bos taurus (Bovine). Detection Range: 39 ng/mL-2500 ng/mL. Sensitivity: 39 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5209