Learn More
Biomatik Corporation Goat Vitamin B12 (Vb12) Elisa Kit
Supplier:  Biomatik Corporation EKC42764
Encompass_Preferred
Goat Vitamin B12 (Vb12) Elisa Kit for detecting Vitamin B12 (Vb12) in serum, plasma, tissue homogenates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5220