Learn More
Biomatik Corporation Mouse glucoprotein 130,gp130 ELISA KIT
Supplier:  Biomatik Corporation EKC42810
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/ml -2000 pg/ml. Sensitivity: 7.81 pg/ml. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Not Available. Assay Time: 1-5 hours. UniProt ID: Q00560. Target Synonyms: Il6stInterleukin-6 receptor subunit beta ELISA Kit; IL-6 receptor subunit beta ELISA Kit; IL-6R subunit beta ELISA Kit; IL-6R-beta ELISA Kit; IL-6RB ELISA Kit; Interleukin-6 signal transducer ELISA Kit; Membrane glycoprotein 130 ELISA Kit; gp130 ELISA Kit; Oncostatin-M receptor subunit alpha ELISA Kit; CD antigen CD130 ELISA Kit. Restriction: For Research Use Only.
Catalog No. 50-229-5226