Learn More
Biomatik Corporation Rat Eotaxin 1 (Eotaxin 1/CCL11/ECF)ELISA Kit
Supplier:  Biomatik Corporation EKC42814
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 pg/ml -1000 pg/ml. Sensitivity: 3.9 pg/ml. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Not Available. Assay Time: 1-5 hours. UniProt ID: P97545. Target Synonyms: Ccl11 ELISA Kit; Scya11Eotaxin ELISA Kit; C-C motif chemokine 11 ELISA Kit; Eosinophil chemotactic protein ELISA Kit; Small-inducible cytokine A11 ELISA Kit. Restriction: For Research Use Only.
Catalog No. 50-229-5230