Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily Member 17 (Tnfrsf17) Elisa Kit
Supplier:  Biomatik Corporation EKC42821
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily Member 17 (Tnfrsf17) Elisa Kit for detecting Tumor Necrosis Factor Receptor Superfamily, Member 17 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q02223; TNFRSF17. Target Synonyms: B cell maturation antigen; B cell maturation factor; B cell maturation protein; B-cell maturation protein; BCM; BCMA; CD269; CD269 antigen; TNFRSF17; TNR17_HUMAN; Tumor necrosis factor receptor superfamily member 17. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5237