Learn More
Biomatik Corporation Rat tumor necrosis factor (ligand) superfamily, member 14 (TNFSF14) ELISA kit
Supplier:  Biomatik Corporation EKC42823
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Not Available. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5239