Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Α, Tnf-Α Elisa Kit
Supplier:  Biomatik Corporation EKC42825
Encompass_Preferred
Mouse Tumor Necrosis Factor Α, Tnf-Α Elisa Kit for detecting Tumor Necrosis Factor Α, Tnf-Α in cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06804; Tnf. Target Synonyms: Tnf; Tnfa; Tnfsf2; Tumor necrosis factor; Cachectin; TNF-alpha; Tumor necrosis factor ligand superfamily member 2; TNF-a) [Cleaved into: Tumor necrosis factor; membrane form; N-terminal fragment; NTF); Intracellular domain 1; ICD1); Intracellular domain 2; ICD2); C-domain 1; C-domain 2; Tumor necrosis factor; soluble form]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5241