Learn More
Biomatik Corporation Mouse low density lipoprotein receptor,LDLR ELISA Kit
Supplier:  Biomatik Corporation EKC42829
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312 ng/mL-20 ng/mL. Sensitivity: 0.078 ng/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Not Available. Assay Time: Not Available. UniProt ID: P35951. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5245