Learn More
Biomatik Corporation Human FBLN1 (Fibulin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF57212
Encompass_Preferred
Human FBLN1 (Fibulin-1) ELISA Kit for detecting FBLN1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.469-30ng/ml. Sensitivity: 0.281ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23142. Target Synonyms: FBLN1; Fibulin-1; FIBL-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5254