Learn More
Biomatik Corporation Human FABP4 (Fatty acid-binding protein, adipocyte) ELISA Kit
Supplier:  Biomatik Corporation EKF57219
Encompass_Preferred
Human FABP4 (Fatty acid-binding protein, adipocyte) ELISA Kit for detecting FABP4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.391-25ng/ml. Sensitivity: 0.234ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15090. Target Synonyms: FABP4; A-FABP; AFABP; ALBP; aP2; Adipocyte lipid-binding protein; Adipocyte-type fatty acid-binding protein; Fatty acid-binding protein 4; fatty acid-binding protein; adipocyte. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5259