Learn More
Biomatik Corporation Human LIPF (Gastric triacylglycerol lipase) ELISA Kit
Supplier:  Biomatik Corporation EKF57228
Encompass_Preferred
Human LIPF (Gastric triacylglycerol lipase) ELISA Kit for detecting LIPF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07098. Target Synonyms: LIPF; Lipase; Gastric; GL; Gastric lipase; Gastric triacylglycerol lipase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5261