Learn More
Biomatik Corporation Human Interferon-induced guanylate-binding protein 1 (GBP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34213
Encompass_Preferred
The Human Interferon-induced guanylate-binding protein 1 (GBP1) ELISA Kit allows for quantitative determination of guanylate binding protein 1, interferon-inducible, 67kDa in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 25 pg/mL-1600 pg/mL. Sensitivity: 6.25 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P32455. Restriction: For Research Use Only.
Catalog No. 50-149-8008