Learn More
Biomatik Corporation Human NMT1 (Glycylpeptide N-tetradecanoyltransferase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57325
Encompass_Preferred
Human NMT1 (Glycylpeptide N-tetradecanoyltransferase 1) ELISA Kit for detecting NMT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 4.688-300pg/ml. Sensitivity: 2.813pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P30419. Target Synonyms: NMT1; Glycylpeptide N-tetradecanoyltransferase 1; NMT; Type I N-myristoyltransferase; Myristoyl-CoA:protein N-myristoyltransferase 1; Peptide N-myristoyltransferase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5293