Learn More
Biomatik Corporation Human CIRBP (Cold-inducible RNA-binding protein) ELISA Kit
Supplier:  Biomatik Corporation EKF57340
Encompass_Preferred
Human CIRBP (Cold-inducible RNA-binding protein) ELISA Kit for detecting CIRBP in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q14011. Target Synonyms: CIRBP; Cold-inducible RNA-binding protein; A18 hnRNP; Glycine-rich RNA-binding protein CIRP; A18HNRNP; CIRP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5300