Learn More
Biomatik Corporation Human NME1 (Nucleoside diphosphate kinase A) ELISA Kit
Supplier:  Biomatik Corporation EKF57352
Encompass_Preferred
Human NME1 (Nucleoside diphosphate kinase A) ELISA Kit for detecting NME1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15531. Target Synonyms: NME1; Nucleoside diphosphate kinase A; NDK A; NDP kinase A; nm23-H1; Tumor metastatic process-associated protein; Granzyme A-activated Dnase; GAAD; Metastasis inhibition factor nm23. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5307