Learn More
Biomatik Corporation Human CFL1 (Cofilin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF57378
Encompass_Preferred
Human CFL1 (Cofilin-1) ELISA Kit for detecting CFL1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23528. Target Synonyms: CFL1; Cofilin 1; Non-Muscle; Cofilin-1; 18 kDa phosphoprotein; p18; Cofilin; non-muscle isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5317