Learn More
Biomatik Corporation Human DEXI (Dexamethasone-induced protein) ELISA Kit
Supplier:  Biomatik Corporation EKF57385
Encompass_Preferred
Human DEXI (Dexamethasone-induced protein) ELISA Kit for detecting DEXI in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O95424. Target Synonyms: DEXI; MYLE; Dexamethasone-induced protein; Protein MYLE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5321