Learn More
Biomatik Corporation Human IL-21 (Interleukin-21) ELISA Kit
Supplier:  Biomatik Corporation EKF57389
Encompass_Preferred
Human IL-21 (Interleukin-21) ELISA Kit for detecting IL-21 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9HBE4. Target Synonyms: IL-21; Interleukin-21; Za11; IL21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5324