Learn More
Biomatik Corporation Human RELB (Transcription factor RelB) ELISA Kit
Supplier:  Biomatik Corporation EKF57393
Encompass_Preferred
Human RELB (Transcription factor RelB) ELISA Kit for detecting RELB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q01201. Target Synonyms: RELB; V-Rel Reticuloendotheliosis Viral Oncogene Homolog B; IREL; I-Rel; nuclear factor of kappalight polypeptide gene enhancer in B-cells 3; v-rel reticuloendotheliosis viral oncogene homolog B; Transcription factor RelB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5326