Learn More
Biomatik Corporation Human NRG-1 (Neuregulin 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57411
Encompass_Preferred
Human NRG-1 (Neuregulin 1) ELISA Kit for detecting NRG-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q02297. Target Synonyms: NRG-1; Neuregulin 1; Heregulin-1; HGL; HRG1-alpha; HRG1-beta 1; NRG1; ARIA; GGF2; GGF; glial growth factor; HGLneu differentiation factor; HRG; HRG1; HRGA; neuregulin 1 type IV beta 1a; MST131; MSTP131; NDFheregulin; alpha; 45kD; ERBB2 p185-activator; neuregulin 1; neuregulin 1 type IV beta 3; pro-neuregulin-1; membrane-bound isoform; pro-NRG1; sensory and motor neuron derived factor; SMDF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5334