Learn More
Biomatik Corporation Human IFI16 (Gamma-interferon-inducible protein 16) ELISA Kit
Supplier:  Biomatik Corporation EKF57417
Encompass_Preferred
Human IFI16 (Gamma-interferon-inducible protein 16) ELISA Kit for detecting IFI16 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q16666. Target Synonyms: IFI16; amma-interferon-inducible protein 16; Ifi-16; Interferon-inducible myeloid differentiation transcriptional activator; IFNGIP1; PYHIN2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5337